Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CWC25
Entrez Gene ID
54883
Gene Name
CWC25 spliceosome associated protein homolog
Gene Aliases
CCDC49
Chromosome Location
Chr.17: 38800434 - 38825350 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017748.4 NP_060218.1 9 - 10 1337 84
NR_073428.1 10 - 11 1454 84
GenBank mRNA AK000298.1 - 9 - 10 1287 84
AK225373.1 - 9 - 10 1307 84
AK296115.1 - 10 - 11 1419 84
BC008833.2 - 9 - 10 1323 84
CR749362.1 - 5 - 6 519 84

View Full Product Details