Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRGPRX3
Entrez Gene ID
117195
Gene Name
MAS related GPR family member X3
Gene Aliases
GPCR, MRGX3, SNSR1, SNSR2
Chromosome Location
Chr.11: 18120955 - 18138480 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054031.3 NP_473372.3 3 - 3 1090 82
XM_011519882.2 XP_011518184.1 2 - 2 945 82
GenBank mRNA BC067292.1 - 3 - 3 1090 82
EU432126.1 - 1 - 1 729 82

View Full Product Details