Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PALM2
Entrez Gene ID
114299
Gene Name
paralemmin 2
Gene Aliases
AKAP2
Chromosome Location
Chr.9: 109640788 - 109951476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037293.2 NP_001032370.1 1 - 2 79 78
GenBank mRNA AK095003.1 - 1 - 2 75 78
BC039306.1 - 1 - 2 81 78

View Full Product Details