The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014671.2 | NP_055486.2 | 21 - 22 | 3258 | 97 |
XM_005249564.4 | XP_005249621.1 | 20 - 21 | 3192 | 97 | |
XM_017012818.1 | XP_016868307.1 | 22 - 23 | 3530 | 97 | |
GenBank mRNA | AK127280.1 | - | 6 - 7 | 3529 | 97 |
AK295138.1 | - | 21 - 22 | 2938 | 97 | |
D13635.1 | - | 21 - 22 | 3249 | 97 |