The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014671.2 | NP_055486.2 | 6 - 7 | 925 | 97 |
XM_005249564.4 | XP_005249621.1 | 5 - 6 | 859 | 97 | |
XM_017012818.1 | XP_016868307.1 | 7 - 8 | 1197 | 97 | |
GenBank mRNA | AK295138.1 | - | 6 - 7 | 605 | 97 |
BC014029.2 | - | 7 - 8 | 685 | 97 | |
BC026241.1 | - | 5 - 6 | 800 | 97 | |
D13635.1 | - | 6 - 7 | 916 | 97 |