The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014671.2 | NP_055486.2 | 7 - 8 | 1081 | 158 |
XM_005249564.4 | XP_005249621.1 | 6 - 7 | 1015 | 158 | |
XM_017012818.1 | XP_016868307.1 | 8 - 9 | 1353 | 158 | |
GenBank mRNA | AK295138.1 | - | 7 - 8 | 761 | 158 |
BC014029.2 | - | 8 - 9 | 841 | 158 | |
BC026241.1 | - | 6 - 7 | 956 | 158 | |
D13635.1 | - | 7 - 8 | 1072 | 158 |