The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253.3 | NP_001244.1 | 6 - 7 | 1070 | 143 |
GenBank mRNA | AB007892.2 | - | 6 - 7 | 842 | 143 |
AK128737.1 | - | 6 - 7 | 878 | 143 | |
AK299738.1 | - | 6 - 7 | 797 | 143 | |
BC001568.1 | - | 6 - 7 | 859 | 143 | |
U86753.1 | - | 6 - 7 | 760 | 143 |