Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKIV2L2
Entrez Gene ID
23517
Gene Name
Ski2 like RNA helicase 2
Gene Aliases
Dob1, KIAA0052, Mtr4, fSAP118
Chromosome Location
Chr.5: 55307748 - 55425581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015360.4 NP_056175.3 1 - 2 403 165
GenBank mRNA AK291649.1 - 1 - 2 150 165
BC028604.2 - 1 - 2 161 165
BC065258.1 - 1 - 2 142 165
BC104996.1 - 1 - 2 182 165
BC113509.1 - 1 - 2 182 165
BX640789.1 - 1 - 2 363 165
D29641.2 - 1 - 2 149 165
DA524147.1 - 1 - 2 404 165

View Full Product Details