The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006209.4 | NP_006200.3 | 12 - 13 | 1213 | 70 |
XM_006716584.1 | XP_006716647.1 | 12 - 13 | 1191 | 70 | |
XM_006716585.1 | XP_006716648.1 | 12 - 13 | 1191 | 70 | |
XM_006716587.1 | XP_006716650.1 | 12 - 13 | 1191 | 70 | |
GenBank mRNA | L35594.1 | - | 12 - 13 | 1176 | 70 |