Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPC1L1
Entrez Gene ID
29881
Gene Name
NPC1 like intracellular cholesterol transporter 1
Gene Aliases
NPC11L1
Chromosome Location
Chr.7: 44512440 - 44541650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101648.1 NP_001095118.1 4 - 5 1910 59
NM_001300967.1 NP_001287896.1 4 - 5 1910 59
NM_013389.2 NP_037521.2 4 - 5 1910 59
XM_011515326.2 XP_011513628.1 4 - 5 1956 59
XM_011515327.2 XP_011513629.1 4 - 5 2245 59
GenBank mRNA AF192522.1 - 4 - 5 1910 59
AF192523.1 - 4 - 5 1910 59
AY437865.1 - 4 - 5 1854 59
AY515256.1 - 4 - 5 1854 59
BC117178.1 - 4 - 5 1908 59
BC143756.1 - 4 - 5 1908 59
FJ481111.1 - 4 - 5 1854 59

View Full Product Details