The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018711.4 | NP_061181.1 | 11 - 12 | 1418 | 63 |
GenBank mRNA | AK309780.1 | - | 2 - 3 | 163 | 63 |
BC033587.1 | - | 11 - 12 | 1218 | 63 | |
BC094722.1 | - | 11 - 12 | 1065 | 63 |