Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178075.1 NP_001171546.1 5 - 6 848 73
NM_001178076.1 NP_001171547.1 3 - 4 614 73
NM_001178077.1 NP_001171548.1 3 - 4 547 73
NM_001673.4 NP_001664.3 5 - 6 922 73
NM_133436.3 NP_597680.2 5 - 6 1177 73
NM_183356.3 NP_899199.2 6 - 7 1200 73
GenBank mRNA AK000379.1 - 5 - 6 1145 73
AK302189.1 - 5 - 6 742 73
AK302242.1 - 3 - 4 495 73
AK316224.1 - 3 - 4 549 73
BC008723.2 - 6 - 7 1027 73
BC014621.2 - 5 - 6 798 73
BT007113.1 - 3 - 4 671 73
M15798.1 - 4 - 5 718 73
M27396.1 - 5 - 6 849 73

View Full Product Details