Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178075.1 NP_001171546.1 6 - 7 954 71
NM_001178076.1 NP_001171547.1 4 - 5 720 71
NM_001178077.1 NP_001171548.1 4 - 5 653 71
NM_001673.4 NP_001664.3 6 - 7 1028 71
NM_133436.3 NP_597680.2 6 - 7 1283 71
NM_183356.3 NP_899199.2 7 - 8 1306 71
GenBank mRNA AK302189.1 - 6 - 7 848 71
AK302242.1 - 4 - 5 601 71
AK316224.1 - 4 - 5 655 71
BC008723.2 - 7 - 8 1133 71
BC014621.2 - 6 - 7 904 71
BT007113.1 - 4 - 5 777 71
M15798.1 - 5 - 6 824 71
M27396.1 - 6 - 7 955 71

View Full Product Details