Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG1
Entrez Gene ID
57480
Gene Name
pleckstrin homology and RhoGEF domain containing G1
Gene Aliases
ARHGEF41
Chromosome Location
Chr.6: 150599863 - 150843665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029884.1 NP_001025055.1 2 - 3 215 72
XM_005267065.3 XP_005267122.1 3 - 4 241 72
XM_006715521.1 XP_006715584.1 3 - 4 468 72
XM_011535983.2 XP_011534285.1 2 - 3 1516 72

View Full Product Details