Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RCC1
- Entrez Gene ID
-
1104
- Gene Name
- regulator of chromosome condensation 1
- Gene Aliases
-
CHC1, RCC1-I, SNHG3-RCC1
- Chromosome Location
-
Chr.1: 28505943 - 28539196 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_030726.1 |
|
3 - 4 |
252 |
97 |
View Full Product Details