The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048194.2 | NP_001041659.1 | 3 - 4 | 251 | 67 |
GenBank mRNA | BC069198.1 | - | 3 - 4 | 216 | 67 |
KU177975.1 | - | 2 - 3 | 166 | 67 |