Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTOV1
Entrez Gene ID
53635
Gene Name
prostate tumor overexpressed 1
Gene Aliases
ACID2, PTOV-1
Chromosome Location
Chr.19: 49850726 - 49860744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305105.1 NP_001292034.1 8 - 9 1087 118
NM_001305108.1 NP_001292037.1 8 - 9 928 118
NM_017432.4 NP_059128.2 8 - 9 1087 118
NR_130963.1 8 - 9 923 118
XM_005258998.4 XP_005259055.1 8 - 9 1078 118
XM_006723244.3 XP_006723307.1 8 - 9 1078 118
XM_011527033.1 XP_011525335.1 8 - 9 913 118
XM_011527034.2 XP_011525336.1 8 - 9 1078 118
XM_011527035.1 XP_011525337.1 8 - 9 1037 118
XM_017026879.1 XP_016882368.1 8 - 9 1078 118
XM_017026880.1 XP_016882369.1 8 - 9 907 118
GenBank mRNA AF218026.1 - 3 - 4 1158 118
AJ617480.1 - 8 - 9 1016 118
AK057817.1 - 8 - 9 923 118
AY358168.1 - 8 - 9 909 118
BC015172.1 - 2 - 3 102 118
BC042921.1 - 8 - 9 982 118
U79287.1 - 8 - 9 820 118

View Full Product Details