Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSUH2
Entrez Gene ID
51066
Gene Name
ssu-2 homolog (C. elegans)
Gene Aliases
C3orf32, SSU-2, fls485
Chromosome Location
Chr.3: 8619400 - 8745035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256748.1 NP_001243677.1 5 - 6 441 104
NM_001256749.1 NP_001243678.1 5 - 6 462 104
NM_015931.2 NP_057015.1 5 - 6 608 104
NR_046358.1 6 - 7 689 104
XM_011533774.1 XP_011532076.1 13 - 14 2157 104
XM_017006510.1 XP_016861999.1 6 - 7 4922 104
XM_017006511.1 XP_016862000.1 12 - 13 1845 104
XM_017006512.1 XP_016862001.1 12 - 13 2014 104
XM_017006513.1 XP_016862002.1 11 - 12 1850 104
XM_017006514.1 XP_016862003.1 11 - 12 1756 104
XM_017006515.1 XP_016862004.1 13 - 14 2911 104
XM_017006516.1 XP_016862005.1 8 - 9 1272 104
XM_017006517.1 XP_016862006.1 8 - 9 1225 104
XM_017006518.1 XP_016862007.1 13 - 14 2136 104
XM_017006519.1 XP_016862008.1 6 - 7 4867 104
XM_017006520.1 XP_016862009.1 6 - 7 1218 104
XM_017006521.1 XP_016862010.1 9 - 10 1376 104
XM_017006522.1 XP_016862011.1 9 - 10 1267 104
XM_017006523.1 XP_016862012.1 7 - 8 970 104
XM_017006524.1 XP_016862013.1 8 - 9 1208 104
XM_017006525.1 XP_016862014.1 11 - 12 1777 104
XM_017006526.1 XP_016862015.1 6 - 7 856 104
XM_017006527.1 XP_016862016.1 12 - 13 1899 104
XM_017006528.1 XP_016862017.1 6 - 7 895 104
XM_017006529.1 XP_016862018.1 6 - 7 925 104
XM_017006530.1 XP_016862019.1 8 - 9 1358 104
XM_017006531.1 XP_016862020.1 7 - 8 1280 104
XM_017006532.1 XP_016862021.1 6 - 7 5780 104
XM_017006533.1 XP_016862022.1 12 - 13 1932 104
XM_017006534.1 XP_016862023.1 12 - 13 1878 104
XM_017006535.1 XP_016862024.1 6 - 7 6056 104
XM_017006536.1 XP_016862025.1 6 - 7 6059 104
XM_017006537.1 XP_016862026.1 6 - 7 6059 104
XM_017006538.1 XP_016862027.1 10 - 11 1475 104
XM_017006539.1 XP_016862028.1 5 - 6 587 104
XM_017006540.1 XP_016862029.1 6 - 7 766 104
XM_017006541.1 XP_016862030.1 3 - 4 408 104
XM_017006542.1 XP_016862031.1 3 - 4 386 104
GenBank mRNA AB024705.1 - 5 - 6 581 104
AK057972.1 - 6 - 7 685 104
AK093066.1 - 11 - 12 1638 104
AK122938.1 - 10 - 11 1423 104
AK225925.1 - 6 - 7 685 104
AK300175.1 - 5 - 6 441 104
BC052614.1 - 5 - 6 445 104
DB010779.1 - 5 - 6 462 104

View Full Product Details