Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
Chromosome Location
Chr.3: 107522936 - 107811329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142568.2 NP_001136040.1 9 - 10 1147 86
NM_001276286.1 NP_001263215.1 9 - 10 1147 86
NM_020235.6 NP_064620.2 9 - 10 1147 86
XM_005247642.3 XP_005247699.1 9 - 10 1063 86
XM_005247643.3 XP_005247700.1 9 - 10 1075 86
XM_005247644.3 XP_005247701.1 10 - 11 1238 86
XM_011512999.2 XP_011511301.1 9 - 10 1147 86
XM_011513000.1 XP_011511302.1 9 - 10 1275 86
XM_011513001.1 XP_011511303.1 9 - 10 1115 86
XM_011513002.2 XP_011511304.1 9 - 10 1147 86
XM_011513003.2 XP_011511305.1 9 - 10 1147 86
XM_011513004.2 XP_011511306.1 9 - 10 1275 86
XM_011513005.2 XP_011511307.1 9 - 10 1147 86
XM_011513006.2 XP_011511308.1 9 - 10 1147 86
XM_011513007.2 XP_011511309.1 9 - 10 1147 86
XM_011513010.2 XP_011511312.1 9 - 10 1147 86
XM_017006881.1 XP_016862370.1 11 - 12 1288 86
XM_017006882.1 XP_016862371.1 10 - 11 1238 86
XM_017006883.1 XP_016862372.1 9 - 10 1147 86
GenBank mRNA AF161457.1 - 9 - 10 1127 86
AF276948.1 - 6 - 7 845 86
AF276949.1 - 9 - 10 1078 86
AF283775.2 - 9 - 10 1141 86
AF454941.1 - 9 - 10 1140 86
AF454942.1 - 7 - 8 1020 86
AK057010.1 - 9 - 10 1093 86
AK291757.1 - 9 - 10 1093 86
AK307668.1 - 8 - 9 1187 86
AK314434.1 - 9 - 10 1098 86
AL136769.1 - 7 - 8 1020 86
BC110903.1 - 7 - 8 1057 86
BC131718.1 - 9 - 10 1040 86

View Full Product Details