The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009813.2 | NP_001009813.1 | 10 - 11 | 1393 | 141 |
NM_001301059.1 | NP_001287988.1 | 10 - 11 | 1444 | 141 | |
NM_020160.2 | NP_064545.1 | 10 - 11 | 1582 | 141 | |
XM_011527136.2 | XP_011525438.1 | 10 - 11 | 1591 | 141 | |
XM_011527138.2 | XP_011525440.1 | 8 - 9 | 1441 | 141 | |
XM_011527139.2 | XP_011525441.1 | 10 - 11 | 1112 | 141 | |
XM_011527140.1 | XP_011525442.1 | 10 - 11 | 1611 | 141 | |
XM_017027011.1 | XP_016882500.1 | 10 - 11 | 1642 | 141 | |
GenBank mRNA | AB209455.1 | - | 5 - 6 | 622 | 141 |
AK098710.1 | - | 10 - 11 | 1306 | 141 | |
AK289950.1 | - | 10 - 11 | 1176 | 141 | |
AY091593.1 | - | 8 - 9 | 673 | 141 | |
BC025404.1 | - | 10 - 11 | 1345 | 141 | |
BC045741.1 | - | 3 - 4 | 141 | ||
BC069251.1 | - | 10 - 11 | 1015 | 141 | |
BI522513.1 | - | 3 - 4 | 237 | 141 |