The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017643.2 | NP_060113.2 | 15 - 16 | 2029 | 112 |
XM_005257467.4 | XP_005257524.1 | 14 - 15 | 1974 | 112 | |
XM_005257468.4 | XP_005257525.1 | 14 - 15 | 1967 | 112 | |
XM_011524922.2 | XP_011523224.1 | 14 - 15 | 1976 | 112 | |
XM_011524923.2 | XP_011523225.1 | 14 - 15 | 1973 | 112 | |
XM_011524924.2 | XP_011523226.1 | 14 - 15 | 1969 | 112 | |
XM_011524925.1 | XP_011523227.1 | 15 - 16 | 2090 | 112 | |
XM_011524926.2 | XP_011523228.1 | 15 - 16 | 2194 | 112 | |
XM_011524928.2 | XP_011523230.1 | 14 - 15 | 2131 | 112 | |
XM_011524929.2 | XP_011523231.1 | 15 - 16 | 2045 | 112 | |
XM_011524930.1 | XP_011523232.1 | 14 - 15 | 1767 | 112 | |
XM_011524931.2 | XP_011523233.1 | 13 - 14 | 1601 | 112 | |
XM_011524933.2 | XP_011523235.1 | 16 - 17 | 2151 | 112 | |
XM_017024782.1 | XP_016880271.1 | 15 - 16 | 2341 | 112 | |
GenBank mRNA | CN310792.1 | - | 1 - 2 | 61 | 112 |