Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MBTD1
Entrez Gene ID
54799
Gene Name
mbt domain containing 1
Gene Aliases
SA49P01
Chromosome Location
Chr.17: 51177419 - 51260511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017643.2 NP_060113.2 12 - 13 1569 119
XM_005257467.4 XP_005257524.1 11 - 12 1514 119
XM_005257468.4 XP_005257525.1 11 - 12 1507 119
XM_011524922.2 XP_011523224.1 11 - 12 1516 119
XM_011524923.2 XP_011523225.1 11 - 12 1513 119
XM_011524924.2 XP_011523226.1 11 - 12 1509 119
XM_011524925.1 XP_011523227.1 12 - 13 1630 119
XM_011524926.2 XP_011523228.1 12 - 13 1734 119
XM_011524928.2 XP_011523230.1 11 - 12 1671 119
XM_011524929.2 XP_011523231.1 12 - 13 1585 119
XM_011524930.1 XP_011523232.1 11 - 12 1307 119
XM_011524931.2 XP_011523233.1 10 - 11 1141 119
XM_011524933.2 XP_011523235.1 13 - 14 1691 119
XM_017024782.1 XP_016880271.1 12 - 13 1881 119
GenBank mRNA AK000062.1 - 8 - 9 901 119
BC034364.1 - 12 - 13 1540 119
BC101736.1 - 7 - 8 830 119

View Full Product Details