The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004245.3 | NP_004236.1 | 11 - 12 | 1766 | 83 |
NM_201631.3 | NP_963925.2 | 12 - 13 | 2012 | 83 | |
XM_011522230.2 | XP_011520532.1 | 6 - 7 | 1194 | 83 | |
XM_017022729.1 | XP_016878218.1 | 6 - 7 | 1218 | 83 | |
GenBank mRNA | AF035960.1 | - | 12 - 13 | 2012 | 83 |
AF035961.1 | - | 11 - 12 | 1766 | 83 | |
AK298479.1 | - | 8 - 9 | 1428 | 83 | |
BC119009.1 | - | 12 - 13 | 2012 | 83 | |
BC122859.1 | - | 12 - 13 | 2012 | 83 |