Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSUH2
Entrez Gene ID
51066
Gene Name
ssu-2 homolog (C. elegans)
Gene Aliases
C3orf32, SSU-2, fls485
Chromosome Location
Chr.3: 8619400 - 8745035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006525.1 XP_016862014.1 6 - 7 955 83
XM_017006538.1 XP_016862027.1 6 - 7 955 83
GenBank mRNA AK093066.1 - 7 - 8 1118 83

View Full Product Details