Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR3
Entrez Gene ID
8897
Gene Name
myotubularin related protein 3
Gene Aliases
FYVE-DSP1, ZFYVE10
Chromosome Location
Chr.22: 29883165 - 30030868 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153050.2 NP_694690.1 18 - 19 3640 65
XM_005261803.2 XP_005261860.1 20 - 21 3869 65
XM_005261804.3 XP_005261861.1 19 - 20 3786 65
XM_005261806.2 XP_005261863.1 19 - 20 3751 65
XM_017029028.1 XP_016884517.1 18 - 19 3643 65
XM_017029030.1 XP_016884519.1 17 - 18 3538 65
XM_017029033.1 XP_016884522.1 13 - 14 3070 65
XM_017029034.1 XP_016884523.1 19 - 20 5073 65
GenBank mRNA AF233436.2 - 15 - 16 3321 65

View Full Product Details