The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021090.3 | NP_066576.1 | 3 - 4 | 323 | 85 |
NM_153050.2 | NP_694690.1 | 3 - 4 | 323 | 85 | |
NM_153051.2 | NP_694691.1 | 3 - 4 | 323 | 85 | |
XM_005261803.2 | XP_005261860.1 | 3 - 4 | 323 | 85 | |
XM_005261805.2 | XP_005261862.1 | 3 - 4 | 323 | 85 | |
XM_005261806.2 | XP_005261863.1 | 3 - 4 | 323 | 85 | |
XM_005261808.2 | XP_005261865.1 | 3 - 4 | 323 | 85 | |
XM_017029032.1 | XP_016884521.1 | 3 - 4 | 206 | 85 | |
XM_017029034.1 | XP_016884523.1 | 3 - 4 | 296 | 85 | |
GenBank mRNA | AB002369.1 | - | 3 - 4 | 242 | 85 |
AK289581.1 | - | 3 - 4 | 323 | 85 | |
BC142713.1 | - | 3 - 4 | 279 | 85 | |
BC148216.1 | - | 2 - 3 | 142 | 85 | |
BC151217.1 | - | 3 - 4 | 337 | 85 | |
BC152455.1 | - | 3 - 4 | 242 | 85 | |
DA126318.1 | - | 3 - 4 | 323 | 85 |