The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195483.1 | NP_001182412.1 | 10 - 11 | 1853 | 149 |
NM_024628.5 | NP_078904.3 | 11 - 12 | 1914 | 149 | |
GenBank mRNA | AF345197.1 | - | 10 - 11 | 1803 | 149 |
AF390442.1 | - | 10 - 11 | 1854 | 149 | |
AK026841.1 | - | 7 - 8 | 1250 | 149 | |
AK308484.1 | - | 6 - 7 | 1171 | 149 | |
BC020506.1 | - | 3 - 4 | 816 | 149 | |
BC126158.1 | - | 10 - 11 | 1824 | 149 | |
BC126160.1 | - | 10 - 11 | 1824 | 149 | |
BX538023.1 | - | 5 - 6 | 1237 | 149 | |
CR457363.1 | - | 2 - 3 | 585 | 149 |