The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256789.2 | NP_001243718.1 | 29 - 30 | 3662 | 69 |
NM_001256790.2 | NP_001243719.1 | 29 - 30 | 3500 | 69 | |
NM_005183.3 | NP_005174.2 | 29 - 30 | 3695 | 69 | |
XM_011543983.2 | XP_011542285.1 | 29 - 30 | 3533 | 69 | |
XM_017029836.1 | XP_016885325.1 | 9 - 10 | 952 | 69 | |
GenBank mRNA | AF067227.1 | - | 29 - 30 | 3438 | 69 |
AF201304.1 | - | 29 - 30 | 3695 | 69 | |
AJ224874.1 | - | 29 - 30 | 3662 | 69 | |
JF701915.1 | - | 29 - 30 | 3600 | 69 |