The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003878.2 | NP_003869.1 | 7 - 8 | 973 | 139 |
GenBank mRNA | AK290450.1 | - | 7 - 8 | 717 | 139 |
BC025025.1 | - | 7 - 8 | 774 | 139 | |
U55206.1 | - | 7 - 8 | 749 | 139 |