Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr.3: 46436005 - 46485234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199149.1 NP_001186078.1 11 - 12 1543 94
NM_001321121.1 NP_001308050.1 11 - 12 1648 94
NM_001321122.1 NP_001308051.1 14 - 15 1887 94
NM_002343.5 NP_002334.2 11 - 12 1654 94
GenBank mRNA AF332168.1 - 11 - 12 1361 94
AK093852.1 - 14 - 15 1890 94
AK290859.1 - 11 - 12 1399 94
AK292813.1 - 11 - 12 1399 94
AK298035.1 - 11 - 12 1543 94
AK303995.1 - 3 - 4 262 94
AK316330.1 - 11 - 12 1463 94
AY137470.1 - 11 - 12 1361 94
AY165046.1 - 11 - 12 1373 94
AY178998.2 - 11 - 12 1361 94
AY360320.1 - 11 - 12 1356 94
AY493417.1 - 11 - 12 1471 94
AY875691.1 - 11 - 12 1361 94
BC022347.1 - 11 - 12 1399 94
EU779935.1 - 11 - 12 1361 94
EU794669.1 - 11 - 12 1396 94
KF992208.1 - 11 - 12 1373 94
KT006756.1 - 11 - 12 1361 94
M18642.1 - 1 - 2 56 94
M83202.1 - 11 - 12 1368 94
M83205.1 - 11 - 12 1347 94
M93150.1 - 11 - 12 1378 94
U07643.1 - 11 - 12 1458 94
X52941.1 - 11 - 12 1355 94
X53961.1 - 11 - 12 1655 94

View Full Product Details