The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014496.4 | NP_055311.1 | 19 - 20 | 1887 | 108 |
XM_011530917.2 | XP_011529219.1 | 22 - 23 | 2187 | 108 | |
XM_011530918.2 | XP_011529220.1 | 21 - 22 | 2099 | 108 | |
XM_011530919.2 | XP_011529221.1 | 20 - 21 | 2025 | 108 | |
XM_011530920.2 | XP_011529222.1 | 18 - 19 | 1836 | 108 | |
XM_017029423.1 | XP_016884912.1 | 19 - 20 | 3057 | 108 | |
XM_017029424.1 | XP_016884913.1 | 21 - 22 | 2753 | 108 | |
XM_017029425.1 | XP_016884914.1 | 20 - 21 | 2601 | 108 | |
XM_017029426.1 | XP_016884915.1 | 19 - 20 | 1946 | 108 | |
GenBank mRNA | AF184965.1 | - | 19 - 20 | 1931 | 108 |
AK313240.1 | - | 19 - 20 | 1866 | 108 | |
BC143647.1 | - | 20 - 21 | 2025 | 108 | |
BC143648.1 | - | 21 - 22 | 2099 | 108 | |
CR536566.1 | - | 19 - 20 | 1851 | 108 |