Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
Chromosome Location
Chr.5: 181236928 - 181243906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006098.4 NP_006089.1 4 - 5 625 75
GenBank mRNA AK095666.1 - 4 - 5 497 75
AK222488.1 - 4 - 5 625 75
AK301029.1 - 4 - 5 461 75
AK301468.1 - 4 - 5 551 75
AY336089.1 - 4 - 5 526 75
BC000214.1 - 4 - 5 643 75
BC000366.2 - 4 - 5 613 75
BC010119.2 - 4 - 5 600 75
BC014256.1 - 4 - 5 602 75
BC014788.1 - 4 - 5 609 75
BC017287.2 - 4 - 5 590 75
BC019093.2 - 4 - 5 603 75
BC019362.1 - 4 - 5 607 75
BC021993.1 - 4 - 5 625 75
BC029996.1 - 4 - 5 452 75
BC032006.1 - 4 - 5 604 75
BC035460.1 - 4 - 5 606 75
CR456978.1 - 4 - 5 519 75
CR541909.1 - 4 - 5 519 75
GU727632.1 - 4 - 5 519 75
M24194.1 - 4 - 5 614 75

View Full Product Details