The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 6 - 7 | 935 | 66 |
XM_006723769.2 | XP_006723832.1 | 6 - 7 | 944 | 66 | |
GenBank mRNA | AB019219.1 | - | 6 - 7 | 849 | 66 |
AF026031.1 | - | 6 - 7 | 866 | 66 | |
AF221842.1 | - | 6 - 7 | 872 | 66 | |
AK001554.1 | - | 6 - 7 | 878 | 66 | |
AK225993.1 | - | 6 - 7 | 889 | 66 | |
AK314310.1 | - | 6 - 7 | 859 | 66 | |
BC001666.2 | - | 6 - 7 | 813 | 66 |