The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 7 - 8 | 1027 | 76 |
XM_006723769.2 | XP_006723832.1 | 7 - 8 | 1036 | 76 | |
GenBank mRNA | AB019219.1 | - | 7 - 8 | 941 | 76 |
AF026031.1 | - | 7 - 8 | 958 | 76 | |
AF221842.1 | - | 7 - 8 | 964 | 76 | |
AK225993.1 | - | 7 - 8 | 981 | 76 | |
AK314310.1 | - | 7 - 8 | 951 | 76 | |
BC001666.2 | - | 7 - 8 | 905 | 76 |