Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NID1
Entrez Gene ID
4811
Gene Name
nidogen 1
Gene Aliases
NID
Chromosome Location
Chr.1: 235975830 - 236065181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002508.2 NP_002499.2 12 - 13 2625 79
XM_011544195.2 XP_011542497.1 11 - 12 2496 79
GenBank mRNA AB209448.1 - 12 - 13 2522 79
M27445.1 - 4 - 5 525 79
M30269.1 - 12 - 13 2613 79

View Full Product Details