Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NID1
Entrez Gene ID
4811
Gene Name
nidogen 1
Gene Aliases
NID
Chromosome Location
Chr.1: 235975830 - 236065181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002508.2 NP_002499.2 16 - 17 3326 62
XM_011544195.2 XP_011542497.1 15 - 16 3197 62
GenBank mRNA AB209448.1 - 16 - 17 3208 62
BC045606.1 - 13 - 14 2837 62
CN267119.1 - 2 - 3 276 62
DA269707.1 - 1 - 2 32 62
M27445.1 - 8 - 9 1226 62
M30269.1 - 16 - 17 3314 62

View Full Product Details