The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002508.2 | NP_002499.2 | 8 - 9 | 2089 | 89 |
XM_011544195.2 | XP_011542497.1 | 8 - 9 | 2086 | 89 | |
GenBank mRNA | AB209448.1 | - | 8 - 9 | 1986 | 89 |
AK297234.1 | - | 9 - 10 | 2008 | 89 | |
BC045606.1 | - | 8 - 9 | 1999 | 89 |