Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYST
Entrez Gene ID
1130
Gene Name
lysosomal trafficking regulator
Gene Aliases
CHS, CHS1
Chromosome Location
Chr.1: 235661031 - 235883708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000081.3 NP_000072.2 46 - 47 10749 85
NM_001301365.1 NP_001288294.1 46 - 47 11186 85
XM_011544031.1 XP_011542333.1 47 - 48 10911 85
XM_011544032.1 XP_011542334.1 47 - 48 11314 85
XM_011544033.2 XP_011542335.1 47 - 48 11061 85
XM_011544034.1 XP_011542336.1 46 - 47 10773 85
XM_011544036.2 XP_011542338.1 43 - 44 8565 85
XM_017000150.1 XP_016855639.1 46 - 47 10680 85
GenBank mRNA BX649113.1 - 15 - 16 5559 85

View Full Product Details