Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYST
Entrez Gene ID
1130
Gene Name
lysosomal trafficking regulator
Gene Aliases
CHS, CHS1
Chromosome Location
Chr.1: 235661031 - 235883708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000081.3 NP_000072.2 47 - 48 10886 84
NM_001301365.1 NP_001288294.1 47 - 48 11323 84
XM_011544031.1 XP_011542333.1 48 - 49 11048 84
XM_011544032.1 XP_011542334.1 48 - 49 11451 84
XM_011544033.2 XP_011542335.1 48 - 49 11198 84
XM_011544034.1 XP_011542336.1 47 - 48 10910 84
XM_011544036.2 XP_011542338.1 44 - 45 8702 84
XM_017000150.1 XP_016855639.1 47 - 48 10817 84
GenBank mRNA BX649113.1 - 16 - 17 5696 84
U67615.1 - 47 - 48 10893 84

View Full Product Details