The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195010.1 | NP_001181939.1 | 13 - 14 | 1715 | 77 |
NM_021180.3 | NP_067003.2 | 13 - 14 | 1611 | 77 | |
NM_198173.2 | NP_937816.1 | 13 - 14 | 1780 | 77 | |
NM_198174.2 | NP_937817.3 | 13 - 14 | 1780 | 77 | |
XM_011541869.1 | XP_011540171.1 | 13 - 14 | 1661 | 77 | |
XM_011541870.2 | XP_011540172.1 | 12 - 13 | 1582 | 77 | |
GenBank mRNA | AK074386.1 | - | 13 - 14 | 1677 | 77 |
AK315164.1 | - | 13 - 14 | 1582 | 77 | |
AL137763.1 | - | 1 - 2 | 96 | 77 | |
AY231160.1 | - | 13 - 14 | 1611 | 77 | |
AY231161.1 | - | 12 - 13 | 1424 | 77 | |
BC036890.2 | - | 13 - 14 | 1726 | 77 |