The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004468.4 | NP_004459.2 | 2 - 3 | 330 | 64 |
GenBank mRNA | AF133732.1 | - | 1 - 2 | 151 | 64 |
AK290641.1 | - | 2 - 3 | 330 | 64 | |
AK309179.1 | - | 2 - 3 | 247 | 64 | |
BC001351.2 | - | 2 - 3 | 320 | 64 | |
BC011697.2 | - | 2 - 3 | 320 | 64 | |
BT007052.1 | - | 1 - 2 | 151 | 64 | |
BX384389.2 | - | 2 - 3 | 386 | 64 | |
CR457425.1 | - | 1 - 2 | 151 | 64 | |
U60116.2 | - | 1 - 2 | 190 | 64 |