The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243878.1 | NP_001230807.1 | 3 - 4 | 505 | 86 |
NM_004468.4 | NP_004459.2 | 4 - 5 | 681 | 86 | |
GenBank mRNA | AF133732.1 | - | 3 - 4 | 502 | 86 |
AK290641.1 | - | 4 - 5 | 681 | 86 | |
BC001351.2 | - | 4 - 5 | 671 | 86 | |
BC011697.2 | - | 4 - 5 | 671 | 86 | |
BC014501.1 | - | 3 - 4 | 492 | 86 | |
BT007052.1 | - | 3 - 4 | 502 | 86 | |
CR457425.1 | - | 3 - 4 | 502 | 86 |