The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001722.2 | NP_001713.2 | 5 - 6 | 574 | 73 |
GenBank mRNA | AK026588.1 | - | 5 - 6 | 636 | 73 |
AY092086.1 | - | 4 - 5 | 491 | 73 | |
BC002603.2 | - | 4 - 5 | 495 | 73 | |
BC004484.2 | - | 4 - 5 | 517 | 73 | |
CR536509.1 | - | 4 - 5 | 491 | 73 | |
CR541803.1 | - | 4 - 5 | 491 | 73 | |
M17754.1 | - | 5 - 6 | 533 | 73 |