The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015878.1 | NP_001015878.1 | 2 - 3 | 293 | 90 |
GenBank mRNA | AB017332.1 | - | 2 - 3 | 298 | 90 |
AF054621.1 | - | 2 - 3 | 298 | 90 |