The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017895.7 | NP_060365.7 | 5 - 6 | 664 | 110 |
XM_011528888.2 | XP_011527190.1 | 5 - 6 | 635 | 110 | |
GenBank mRNA | AF193054.1 | - | 2 - 3 | 221 | 111 |
AK022979.1 | - | 5 - 6 | 534 | 110 | |
AK300135.1 | - | 5 - 6 | 543 | 110 | |
AL832131.1 | - | 5 - 6 | 580 | 110 | |
AY044431.1 | - | 5 - 6 | 603 | 110 | |
BC009304.1 | - | 5 - 6 | 522 | 110 | |
BC011927.2 | - | 5 - 6 | 518 | 110 | |
BC016060.1 | - | 5 - 6 | 518 | 110 | |
BC126287.1 | - | 5 - 6 | 653 | 110 | |
BC130275.1 | - | 5 - 6 | 653 | 110 | |
BC144125.1 | - | 5 - 6 | 653 | 110 | |
EU718675.1 | - | 5 - 6 | 571 | 110 |