Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC9
Entrez Gene ID
80210
Gene Name
armadillo repeat containing 9
Gene Aliases
ARM, KU-MEL-1, NS21
Chromosome Location
Chr.2: 231198546 - 231394991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271466.2 NP_001258395.1 13 - 14 1369 88
NM_001291656.1 NP_001278585.1 14 - 15 1522 88
NM_025139.5 NP_079415.3 14 - 15 1524 88
XM_011511905.1 XP_011510207.1 14 - 15 1442 88
XM_011511906.1 XP_011510208.1 14 - 15 1442 88
XM_011511907.1 XP_011510209.1 13 - 14 1343 88
XM_011511908.2 XP_011510210.1 14 - 15 1442 88
XM_011511909.2 XP_011510211.1 14 - 15 1442 88
XM_011511910.1 XP_011510212.1 14 - 15 1442 88
XM_011511911.1 XP_011510213.1 14 - 15 1442 88
XM_011511912.1 XP_011510214.1 14 - 15 1442 88
XM_011511913.2 XP_011510215.1 14 - 15 1442 88
XM_011511914.2 XP_011510216.1 14 - 15 1442 88
XM_011511915.2 XP_011510217.1 14 - 15 1442 88
XM_011511916.2 XP_011510218.1 14 - 15 1442 88
XM_011511919.2 XP_011510221.1 4 - 5 830 88
XM_017005018.1 XP_016860507.1 14 - 15 1700 88
XM_017005019.1 XP_016860508.1 14 - 15 1618 88
XM_017005020.1 XP_016860509.1 14 - 15 1700 88
XM_017005021.1 XP_016860510.1 14 - 15 1442 88
XM_017005022.1 XP_016860511.1 14 - 15 1442 88
XM_017005023.1 XP_016860512.1 14 - 15 1700 88
XM_017005024.1 XP_016860513.1 13 - 14 1343 88
XM_017005025.1 XP_016860514.1 14 - 15 1442 88
XM_017005026.1 XP_016860515.1 14 - 15 1641 88
GenBank mRNA AB058771.1 - 3 - 4 265 88
AK022646.1 - 12 - 13 1226 88
AY219922.1 - 14 - 15 1403 88
AY929062.1 - 14 - 15 1396 88
BC004514.2 - 10 - 11 946 88
BC065271.1 - 13 - 14 1356 88
BX537956.1 - 14 - 15 1440 88

View Full Product Details