Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC9
Entrez Gene ID
80210
Gene Name
armadillo repeat containing 9
Gene Aliases
ARM, KU-MEL-1, NS21
Chromosome Location
Chr.2: 231198546 - 231394991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271466.2 NP_001258395.1 5 - 6 635 77
NM_001291656.1 NP_001278585.1 6 - 7 788 77
NM_025139.5 NP_079415.3 6 - 7 790 77
XM_011511905.1 XP_011510207.1 6 - 7 708 77
XM_011511906.1 XP_011510208.1 6 - 7 708 77
XM_011511907.1 XP_011510209.1 6 - 7 708 77
XM_011511908.2 XP_011510210.1 6 - 7 708 77
XM_011511909.2 XP_011510211.1 6 - 7 708 77
XM_011511910.1 XP_011510212.1 6 - 7 708 77
XM_011511911.1 XP_011510213.1 6 - 7 708 77
XM_011511912.1 XP_011510214.1 6 - 7 708 77
XM_011511913.2 XP_011510215.1 6 - 7 708 77
XM_011511914.2 XP_011510216.1 6 - 7 708 77
XM_011511915.2 XP_011510217.1 6 - 7 708 77
XM_011511916.2 XP_011510218.1 6 - 7 708 77
XM_017005018.1 XP_016860507.1 6 - 7 966 77
XM_017005019.1 XP_016860508.1 6 - 7 884 77
XM_017005020.1 XP_016860509.1 6 - 7 966 77
XM_017005021.1 XP_016860510.1 6 - 7 708 77
XM_017005022.1 XP_016860511.1 6 - 7 708 77
XM_017005023.1 XP_016860512.1 6 - 7 966 77
XM_017005024.1 XP_016860513.1 6 - 7 708 77
XM_017005025.1 XP_016860514.1 6 - 7 708 77
XM_017005026.1 XP_016860515.1 6 - 7 907 77
GenBank mRNA AK022646.1 - 4 - 5 492 77
AK309395.1 - - 697 77
AY219922.1 - 6 - 7 669 77
AY929062.1 - 6 - 7 662 77
BC004514.2 - 2 - 3 212 77
BC065271.1 - 5 - 6 622 77
BX537956.1 - 6 - 7 706 77

View Full Product Details