The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324512.1 | NP_001311441.1 | 15 - 16 | 2733 | 74 |
NM_030955.3 | NP_112217.2 | 17 - 18 | 2988 | 74 | |
XM_017009905.1 | XP_016865394.1 | 17 - 18 | 2976 | 74 | |
XM_017009906.1 | XP_016865395.1 | 16 - 17 | 2377 | 74 | |
XM_017009907.1 | XP_016865396.1 | 9 - 10 | 1269 | 74 | |
XM_017009908.1 | XP_016865397.1 | 6 - 7 | 861 | 74 | |
XM_017009909.1 | XP_016865398.1 | 5 - 6 | 775 | 74 | |
GenBank mRNA | AJ250725.1 | - | 17 - 18 | 2975 | 74 |
BC131733.1 | - | 15 - 16 | 2482 | 74 |