Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS12
Entrez Gene ID
81792
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 12
Gene Aliases
PRO4389
Chromosome Location
Chr.5: 33523535 - 33892180 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324512.1 NP_001311441.1 16 - 17 2943 59
NM_030955.3 NP_112217.2 18 - 19 3198 59
GenBank mRNA AJ250725.1 - 18 - 19 3185 59
BC131733.1 - 16 - 17 2692 59

View Full Product Details