The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267571.1 | NP_001254500.1 | 1 - 2 | 549 | 92 |
NM_018421.3 | NP_060891.3 | 1 - 2 | 549 | 92 | |
XM_011518844.2 | XP_011517146.1 | 1 - 2 | 549 | 92 | |
GenBank mRNA | AB209721.1 | - | 1 - 2 | 349 | 92 |
AB449882.1 | - | 1 - 2 | 369 | 92 | |
AF318370.1 | - | 1 - 2 | 408 | 92 | |
AK131290.1 | - | 1 - 2 | 427 | 92 | |
AK298575.1 | - | 1 - 2 | 220 | 92 | |
AY026527.1 | - | 1 - 2 | 461 | 92 |